missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenylbutyronitrile, 97%

$22.68 - $74.34

Chemical Identifiers

CAS 2046-18-6
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00001973
InChI Key ICMVGKQFVMTRLB-UHFFFAOYSA-N
Synonym 4-phenylbutyronitrile, benzenebutanenitrile, 3-cyanopropyl benzene, butyronitrile, 4-phenyl, g-phenylpropylcyanide, 4-phenyl-butyronitrile, acmc-209fai, gamma-phenylbutyronitrile, 1-cyano-3-phenylpropane, d0ft2z
PubChem CID 74897
IUPAC Name 4-phenylbutanenitrile
SMILES C1=CC=C(C=C1)CCCC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0163806 View Documents Alfa Aesar™
L0163806
5g Each for $22.68
 
AAL0163814 View Documents Alfa Aesar™
L0163814
25g Each for $74.34
 
Specifications

Chemical Identifiers

2046-18-6
145.205
ICMVGKQFVMTRLB-UHFFFAOYSA-N
74897
C1=CC=C(C=C1)CCCC#N
C10H11N
MFCD00001973
4-phenylbutyronitrile, benzenebutanenitrile, 3-cyanopropyl benzene, butyronitrile, 4-phenyl, g-phenylpropylcyanide, 4-phenyl-butyronitrile, acmc-209fai, gamma-phenylbutyronitrile, 1-cyano-3-phenylpropane, d0ft2z
4-phenylbutanenitrile

Specifications

105°C (221°F)
5g
C10H11N
UN3276
4-phenylbutyronitrile, benzenebutanenitrile, 3-cyanopropyl benzene, butyronitrile, 4-phenyl, g-phenylpropylcyanide, 4-phenyl-butyronitrile, acmc-209fai, gamma-phenylbutyronitrile, 1-cyano-3-phenylpropane, d0ft2z
C1=CC=C(C=C1)CCCC#N
145.205
145.2
4-Phenylbutyronitrile
129°C to 131°C (10mmHg)
2046-18-6
MFCD00001973
2043002
ICMVGKQFVMTRLB-UHFFFAOYSA-N
4-phenylbutanenitrile
74897
97%
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

TSCA : TSCA