missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Phenylazophenol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC417361000

 View more versions of this product

Catalog No. AC417361000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

198.22
Fine Crystalline Powder
Brown
100.0
4-Hydroxyazobenzene
C12H10N2O
1689-82-3
Glass bottle
≥96.0%
100g
16, 96
96.0
97%
4-Phenylazophenol
MFCD00002330
Authentic

Chemical Identifiers

1689-82-3
100.0
C12H10N2O
MFCD00002330
4-Phenylazophenol
96.0
97%
198.22
4-Hydroxyazobenzene
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.