missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenylazomaleinanil, 95%

$47.18 - $160.38

Chemical Identifiers

CAS 16201-96-0
Molecular Formula C16H11N3O2
Molecular Weight (g/mol) 277.283
MDL Number MFCD00005506
InChI Key DVNPYLMPVFDKGZ-UHFFFAOYSA-N
Synonym 4-phenylazomaleinanil, n-p-phenylazophenyl maleimide, p-phenylazomaleinanil, 1h-pyrrole-2,5-dione, 1-4-phenylazo phenyl, maleimide, n-p-phenylazo phenyl, 1h-pyrrole-2,5-dione, 1-4-phenylazo phenyl, maleimide, n-p-phenylazo phenyl, 1h-pyrrole-2, 1-4-phenylazo phenyl, 1h-pyrrole-2,5-dione, 1-4-2-phenyldiazenyl phenyl, 1h-pyrrole-2,5-dione, 1-4-2-phenyldiazenyl phenyl
PubChem CID 85328
IUPAC Name 1-(4-phenyldiazenylphenyl)pyrrole-2,5-dione
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)N3C(=O)C=CC3=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0087903 View Documents Alfa Aesar™
L0087903
1g Each for $47.18
 
AAL0087906 View Documents Alfa Aesar™
L0087906
5g Each for $160.38
 
Specifications

Chemical Identifiers

16201-96-0
277.283
DVNPYLMPVFDKGZ-UHFFFAOYSA-N
85328
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)N3C(=O)C=CC3=O
C16H11N3O2
MFCD00005506
4-phenylazomaleinanil, n-p-phenylazophenyl maleimide, p-phenylazomaleinanil, 1h-pyrrole-2,5-dione, 1-4-phenylazo phenyl, maleimide, n-p-phenylazo phenyl, 1h-pyrrole-2,5-dione, 1-4-phenylazo phenyl, maleimide, n-p-phenylazo phenyl, 1h-pyrrole-2, 1-4-phenylazo phenyl, 1h-pyrrole-2,5-dione, 1-4-2-phenyldiazenyl phenyl, 1h-pyrrole-2,5-dione, 1-4-2-phenyldiazenyl phenyl
1-(4-phenyldiazenylphenyl)pyrrole-2,5-dione

Specifications

162°C to 165°C
16201-96-0
MFCD00005506
4-phenylazomaleinanil, n-p-phenylazophenyl maleimide, p-phenylazomaleinanil, 1h-pyrrole-2,5-dione, 1-4-phenylazo phenyl, maleimide, n-p-phenylazo phenyl, 1h-pyrrole-2,5-dione, 1-4-phenylazo phenyl, maleimide, n-p-phenylazo phenyl, 1h-pyrrole-2, 1-4-phenylazo phenyl, 1h-pyrrole-2,5-dione, 1-4-2-phenyldiazenyl phenyl, 1h-pyrrole-2,5-dione, 1-4-2-phenyldiazenyl phenyl
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)N3C(=O)C=CC3=O
277.283
277.28
4-Phenylazomaleinanil
1g
C16H11N3O2
236610
DVNPYLMPVFDKGZ-UHFFFAOYSA-N
1-(4-phenyldiazenylphenyl)pyrrole-2,5-dione
85328
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA