Please login to your online account to display your discounted pricing

4-Phenylazobenzoyl Chloride 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  417350010

 View more versions of this product

Catalog No. AC417350010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 244.68
Quantity 1g
Color Orange-red
EINECS Number 203-188-4
TSCA TSCA
Melting Point 93° to 97°C
CAS Max % 100.0
Chemical Name or Material 4-Phenylazobenzoyl chloride
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C13H9ClN2O
MDL Number MFCD00041722
CAS 104-24-5
CAS Min % 97.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Phenylazobenzoyl chloride
Name Note 98%
CAS 104-24-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H9ClN2O
Formula Weight 244.68
MDL Number MFCD00041722