Please login to your online account to display your discounted pricing

4-Phenylazobenzoyl Chloride 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Phenylazobenzoyl chloride
Name Note 98%
CAS 104-24-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H9ClN2O
Formula Weight 244.68
MDL Number MFCD00041722
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC417350010

  SDS
thermo fisher scientific
417350010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 244.68
Quantity 1g
Color Orange-red
EINECS Number 203-188-4
TSCA TSCA
Melting Point 93° to 97°C
CAS Max % 100.0
Chemical Name or Material 4-Phenylazobenzoyl chloride
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C13H9ClN2O
MDL Number MFCD00041722
CAS 104-24-5
CAS Min % 97.5
Infrared Spectrum Authentic