Learn More
4-(Phenylazo)diphenylamine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 212010250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-(Phenylazo)diphenylamine | |
101-75-7 | |
100.0 | |
96% min. (HPLC) | |
C18H15N3 | |
MFCD00003023 | |
16, 314 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether | |
N(C1=CC=CC=C1)C1=CC=C(C=C1)N=NC1=CC=CC=C1 | |
273.34 | |
273.33 |
97% | |
96.0 | |
Authentic | |
Glass bottle | |
C6H5N=NC6H4NHC6H5 | |
25g | |
4-anilinoazobenzene, 4-phenylazo diphenylamine, 4-phenylazodiphenylamine, 4-benzeneazodiphenylamine, benzenamine, n-phenyl-4-phenylazo, azobenzene, 4-anilino, n-phenyl-4-aminoazobenzene, 4-phenylamino azobenzene, diphenylamine, 4-phenylazo, n-phenyl-4-phenylazo aniline | |
VXLFYNFOITWQPM-UHFFFAOYSA-N | |
N-phenyl-4-phenyldiazenylaniline | |
7575 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Warning
EINECSNumber : 202-972-3
RTECSNumber : JK0185000
TSCA : TSCA
RUO â Research Use Only