Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Phenylaminomethyl)benzeneboronic acid pinacol ester, 97%

Chemical Identifiers

Chemical Name or Material 4-(Phenylaminomethyl)benzeneboronic acid pinacol ester
Molecular Formula C19H24BNO2
Formula Weight 309.21
MDL Number MFCD06795652
Synonym 4-(Phenylaminomethyl)phenylboronic acid pinacol ester; N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)]benzyl]aniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2908606

 
alfa aesar™
H2908606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2908603

 
alfa aesar™
H2908603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 309.21
Chemical Name or Material 4-(Phenylaminomethyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C19H24BNO2
MDL Number MFCD06795652
Synonym 4-(Phenylaminomethyl)phenylboronic acid pinacol ester; N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)]benzyl]aniline
Melting Point 114° to 117°C
Quantity 5g