Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Phenylamino)biphenyl, 98%

Chemical Identifiers

Chemical Name or Material 4-(Phenylamino)biphenyl
CAS 32228-99-2
Formula Weight 245.32
MDL Number MFCD01318677
Beilstein 2806457
Synonym N-Phenyl-4-biphenylamine; 4-Phenyldiphenylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6480803

  SDS
alfa aesar™
H6480803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6480806

  SDS
alfa aesar™
H6480806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64808MD

  SDS
alfa aesar™
H64808MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.32
Assay 98%
CAS 32228-99-2
Quantity 1g
Beilstein 2806457
Chemical Name or Material 4-(Phenylamino)biphenyl
Physical Form Solid
Sensitivity Air Sensitive
MDL Number MFCD01318677
Synonym N-Phenyl-4-biphenylamine; 4-Phenyldiphenylamine