Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenyl-3-thiosemicarbazide, 98+%

Chemical Identifiers

Chemical Name or Material 4-Phenyl-3-thiosemicarbazide
CAS 5351-69-9
Molecular Formula C7H9N3S
Formula Weight 167.23
MDL Number MFCD00007615
UN Number UN2811
Beilstein 608285
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1534909

  SDS
alfa aesar™
A1534909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1534918

  SDS
alfa aesar™
A1534918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1534930

  SDS
alfa aesar™
A1534930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.23
Molecular Formula C7H9N3S
RTECS Number VT4025000
MDL Number MFCD00007615
Assay ≥98%
UN Number UN2811
Melting Point 136° to 140°C
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 10g
Chemical Name or Material 4-Phenyl-3-thiosemicarbazide
CAS 5351-69-9
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Beilstein 608285
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.