missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenyl-3-thiosemicarbazide, 98+%

Manufacturer:  Alfa Aesar™ A1534909

 View more versions of this product

Catalog No. AAA1534909



Specifications

Specifications

4-Phenyl-3-thiosemicarbazide
10g
C7H9N3S
UN2811
4-phenyl-3-thiosemicarbazide, 4-phenylthiosemicarbazide, n-phenylhydrazinecarbothioamide, hydrazinecarbothioamide, n-phenyl, phenyl thiosemicarbazide, semicarbazide, 4-phenyl-3-thio, 4-phenyl thiosemicarbazide, usaf el-45, usaf ek-5426, 3-amino-1-phenylthiourea
C1=CC=C(C=C1)NC(=S)NN
167.23
167.23
136°C to 140°C
5351-69-9
MFCD00007615
608285
KKIGUVBJOHCXSP-UHFFFAOYSA-N
1-amino-3-phenylthiourea
730679
≥98%

Chemical Identifiers

5351-69-9
167.23
KKIGUVBJOHCXSP-UHFFFAOYSA-N
730679
C1=CC=C(C=C1)NC(=S)NN
C7H9N3S
MFCD00007615
4-phenyl-3-thiosemicarbazide, 4-phenylthiosemicarbazide, n-phenylhydrazinecarbothioamide, hydrazinecarbothioamide, n-phenyl, phenyl thiosemicarbazide, semicarbazide, 4-phenyl-3-thio, 4-phenyl thiosemicarbazide, usaf el-45, usaf ek-5426, 3-amino-1-phenylthiourea
1-amino-3-phenylthiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : VT4025000

TSCA : TSCA

Documents