missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenyl-2-thiazoleacetonitrile, 97%

$59.26 - $216.39

Chemical Identifiers

CAS 41381-89-9
Molecular Formula C11H8N2S
Molecular Weight (g/mol) 200.259
MDL Number MFCD00551791
InChI Key SOFCSIBVJROWGI-UHFFFAOYSA-N
Synonym 2-4-phenyl-1,3-thiazol-2-yl acetonitrile, 2-4-phenyl-1,3-thiazol-2-yl ethanenitrile, 2-4-phenylthiazol-2-yl acetonitrile, 2-cyanomethyl-4-phenylthiazole, 2-thiazoleacetonitrile, 4-phenyl, 4-phenyl-1,3-thiazol-2-yl acetonitrile, 4-phenyl-2-cyanomethylthiazole, 4-phenyl-thiazol-2-yl-acetonitrile, cambridge id 5156617, maybridge1_006883
PubChem CID 704539
IUPAC Name 2-(4-phenyl-1,3-thiazol-2-yl)acetonitrile
SMILES C1=CC=C(C=C1)C2=CSC(=N2)CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65040MD View Documents Alfa Aesar™
H65040MD
250mg Each for $59.26
 
AAH6504003 View Documents Alfa Aesar™
H6504003
1g Each for $216.39
 
Specifications

Chemical Identifiers

41381-89-9
200.259
SOFCSIBVJROWGI-UHFFFAOYSA-N
704539
C1=CC=C(C=C1)C2=CSC(=N2)CC#N
C11H8N2S
MFCD00551791
2-4-phenyl-1,3-thiazol-2-yl acetonitrile, 2-4-phenyl-1,3-thiazol-2-yl ethanenitrile, 2-4-phenylthiazol-2-yl acetonitrile, 2-cyanomethyl-4-phenylthiazole, 2-thiazoleacetonitrile, 4-phenyl, 4-phenyl-1,3-thiazol-2-yl acetonitrile, 4-phenyl-2-cyanomethylthiazole, 4-phenyl-thiazol-2-yl-acetonitrile, cambridge id 5156617, maybridge1_006883
2-(4-phenyl-1,3-thiazol-2-yl)acetonitrile

Specifications

Brown
41381-89-9
MFCD00551791
2-4-phenyl-1,3-thiazol-2-yl acetonitrile, 2-4-phenyl-1,3-thiazol-2-yl ethanenitrile, 2-4-phenylthiazol-2-yl acetonitrile, 2-cyanomethyl-4-phenylthiazole, 2-thiazoleacetonitrile, 4-phenyl, 4-phenyl-1,3-thiazol-2-yl acetonitrile, 4-phenyl-2-cyanomethylthiazole, 4-phenyl-thiazol-2-yl-acetonitrile, cambridge id 5156617, maybridge1_006883
C1=CC=C(C=C1)C2=CSC(=N2)CC#N
200.259
200.26
97%
250mg
C11H8N2S
UN3439
SOFCSIBVJROWGI-UHFFFAOYSA-N
2-(4-phenyl-1,3-thiazol-2-yl)acetonitrile
704539
Solid
4-Phenyl-2-thiazoleacetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures