missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenyl-2-butanone 95.0+%, TCI America™
Supplier: TCI America B040525ML
Specifications
| 4-Phenyl-2-butanone | |
| Yellow | |
| C10H12O | |
| 25 mL | |
| AKGGYBADQZYZPD-UHFFFAOYSA-N | |
| 4-phenylbutan-2-one | |
| 17355 | |
| ≥95.0% (GC) |
| 2550-26-7 | |
| 115°C | |
| MFCD00008790 | |
| benzylacetone, 4-phenyl-2-butanone, 2-butanone, 4-phenyl, 4-penylbutan-2-one, 1-phenyl-3-butanone, methyl phenethyl ketone, benzyl acetone, methyl 2-phenylethyl ketone, phenethyl methyl ketone, methyl phenylethyl ketone | |
| CC(=O)CCC1=CC=CC=C1 | |
| 148.205 | |
| 148.21 | |
| Liquid |
Chemical Identifiers
| 2550-26-7 | |
| 148.205 | |
| AKGGYBADQZYZPD-UHFFFAOYSA-N | |
| 17355 | |
| CC(=O)CCC1=CC=CC=C1 |
| C10H12O | |
| MFCD00008790 | |
| benzylacetone, 4-phenyl-2-butanone, 2-butanone, 4-phenyl, 4-penylbutan-2-one, 1-phenyl-3-butanone, methyl phenethyl ketone, benzyl acetone, methyl 2-phenylethyl ketone, phenethyl methyl ketone, methyl phenylethyl ketone | |
| 4-phenylbutan-2-one |
Safety and Handling
EINECSNumber : (3)-4474
RTECSNumber : EL9600000
TSCA : Yes