Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenyl-2-(4-pyridyl)thiazole, 97%

$312.19 - $312.19

Chemical Identifiers

Chemical Name or Material 4-Phenyl-2-(4-pyridyl)thiazole
CAS 106950-18-9
Molecular Formula C14H10N2S
Formula Weight 238.31g/mol
MDL Number MFCD02366451
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5175406

 
alfa aesar™
H51754-06
Each for $312.19
Description & Specifications

Specifications

Melting Point 91° to 95°C
Quantity 5g
Chemical Name or Material 4-Phenyl-2-(4-pyridyl)thiazole
CAS 106950-18-9
Assay 97%
Molecular Formula C14H10N2S
Formula Weight 238.31g/mol
MDL Number MFCD02366451
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.