Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenyl-2-(2-pyridyl)thiazole, 97%

$120.97 - $444.85

Chemical Identifiers

Chemical Name or Material 4-Phenyl-2-(2-pyridyl)thiazole
CAS 14384-67-9
Molecular Formula C14H10N2S
Formula Weight 238.31
MDL Number MFCD09058229
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5173103

  SDS
alfa aesar™
H5173103
1g Each for $120.97

AAH5173106

  SDS
alfa aesar™
H5173106
5g Each for $444.85
Description & Specifications

Specifications

Formula Weight 238.31
MDL Number MFCD09058229
Health Hazard 1 Warning
Chemical Name or Material 4-Phenyl-2-(2-pyridyl)thiazole
Assay 97%
Molecular Formula C14H10N2S
CAS 14384-67-9
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 55° to 59°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.