missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenyl-2-(2-pyrazinyl)thiazole-5-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H54637MD

 View more versions of this product

Catalog No. AAH54637MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Phenyl-2-(2-pyrazinyl)thiazole-5-carboxylic acid
1506428-22-3
MFCD20265306
ZCNOYYRPSQWYSP-UHFFFAOYSA-N
4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
73995903
97%
250mg
C14H9N3O2S
4-phenyl-2-2-pyrazinyl thiazole-5-carboxylic acid
C1=CC=C(C=C1)C2=C(SC(=N2)C3=NC=CN=C3)C(=O)O
283.305
283.3

Chemical Identifiers

1506428-22-3
283.305
ZCNOYYRPSQWYSP-UHFFFAOYSA-N
73995903
C1=CC=C(C=C1)C2=C(SC(=N2)C3=NC=CN=C3)C(=O)O
C14H9N3O2S
MFCD20265306
4-phenyl-2-2-pyrazinyl thiazole-5-carboxylic acid
4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents