missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenyl-2-(2-pyrazinyl)thiazole-5-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H5463706

 View more versions of this product

Catalog No. AAH5463706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Phenyl-2-(2-pyrazinyl)thiazole-5-carboxylic acid
1506428-22-3
MFCD20265306
ZCNOYYRPSQWYSP-UHFFFAOYSA-N
4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
73995903
97%
5g
C14H9N3O2S
4-phenyl-2-2-pyrazinyl thiazole-5-carboxylic acid
C1=CC=C(C=C1)C2=C(SC(=N2)C3=NC=CN=C3)C(=O)O
283.305
283.3

Chemical Identifiers

1506428-22-3
283.305
ZCNOYYRPSQWYSP-UHFFFAOYSA-N
73995903
C1=CC=C(C=C1)C2=C(SC(=N2)C3=NC=CN=C3)C(=O)O
C14H9N3O2S
MFCD20265306
4-phenyl-2-2-pyrazinyl thiazole-5-carboxylic acid
4-phenyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents