missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Phenyl-1-butene 98.0+%, TCI America™

Manufacturer:  TCI America P01615ML

 View more versions of this product

Catalog No. P01615ML



Specifications

Specifications

4-Phenyl-1-butene
Colorless
768-56-9
MFCD00026111
PBGVMIDTGGTBFS-UHFFFAOYSA-N
but-3-enylbenzene
13033
Clear Liquid at 20°C
181°C
5mL
C10H12
3295
C=CCCC1=CC=CC=C1
132.206
132.21
≥98.0% (GC)

Chemical Identifiers

768-56-9
132.206
PBGVMIDTGGTBFS-UHFFFAOYSA-N
but-3-enylbenzene
C10H12
MFCD00026111
13033
C=CCCC1=CC=CC=C1
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4520

RTECSNumber : EM9000000

TSCA : No

Documents