missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Phenyl-1-butene 98.0+%, TCI America™
Supplier: TCI America P016125ML
Specifications
4-Phenyl-1-butene | |
Colorless | |
C10H12 | |
25 mL | |
PBGVMIDTGGTBFS-UHFFFAOYSA-N | |
but-3-enylbenzene | |
13033 | |
≥98.0% (GC) |
768-56-9 | |
181°C | |
MFCD00026111 | |
3295 | |
C=CCCC1=CC=CC=C1 | |
132.206 | |
132.21 | |
Liquid |
Chemical Identifiers
768-56-9 | |
132.206 | |
PBGVMIDTGGTBFS-UHFFFAOYSA-N | |
but-3-enylbenzene |
C10H12 | |
MFCD00026111 | |
13033 | |
C=CCCC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (3)-4520
RTECSNumber : EM9000000
TSCA : No