Please login to your online account to display your discounted pricing

4-phenyl-1-buten-4-ol 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC380860050

 View more versions of this product

Catalog No. AC380860050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 148.2
Quantity 5g
Color Clear colorless
Density 0.992g/mL
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.5300 to 1.5320 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C10H12O
Linear Formula C6H5CH(OH)CH2CHCH2
CAS 936-58-3
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 4-Phenyl-1-buten-4-ol
MDL Number MFCD00039617
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Phenyl-1-buten-4-ol
CAS 936-58-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H12O
Linear Formula C6H5CH(OH)CH2CHCH2
Formula Weight 148.2
MDL Number MFCD00039617
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents