missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Phenyl-1-buten-4-ol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5346614

 View more versions of this product

Catalog No. AAH5346614



Description

Description

4-Phenyl-1-buten-4-ol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-1-buten-4-ol is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

4-Phenyl-1-buten-4-ol
0.992
103°C (217°F)
1.53
25 g
RGKVZBXSJFAZRE-UHFFFAOYSA-N
1-phenylbut-3-en-1-ol
220119
97%
936-58-3
234°C
C10H12O
MFCD00039617
1-phenyl-3-buten-1-ol, 4-phenyl-1-buten-4-ol, 1-phenyl-but-3-en-1-ol, acmc-1bdtt, upenn_abs_025, 1-phenyl-3-buten-1-ol #, benzenemethanol,a-2-propen-1-yl, benzenemethanol, .alpha.-2-propenyl
C=CCC(C1=CC=CC=C1)O
148.205
148.2

Chemical Identifiers

936-58-3
148.205
RGKVZBXSJFAZRE-UHFFFAOYSA-N
220119
C=CCC(C1=CC=CC=C1)O
C10H12O
MFCD00039617
1-phenyl-3-buten-1-ol, 4-phenyl-1-buten-4-ol, 1-phenyl-but-3-en-1-ol, acmc-1bdtt, upenn_abs_025, 1-phenyl-3-buten-1-ol #, benzenemethanol,a-2-propen-1-yl, benzenemethanol, .alpha.-2-propenyl
1-phenylbut-3-en-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

P264b-P270-P301+P312-P330-P501c

H302

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only