Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenyl-1-(4-piperidinyl)-4-imidazolin-2-one, 96%

$116.92 - $406.82

Chemical Identifiers

Chemical Name or Material 4-Phenyl-1-(4-piperidinyl)-4-imidazolin-2-one
CAS 205058-28-2
Molecular Formula C14H17N3O
Formula Weight 243.31
MDL Number MFCD08061015
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5082803

  SDS
alfa aesar™
H5082803
1g Each for $406.82

AAH50828MD

  SDS
alfa aesar™
H50828MD
250mg Each for $116.92
Description & Specifications

Specifications

Formula Weight 243.31
Chemical Name or Material 4-Phenyl-1-(4-piperidinyl)-4-imidazolin-2-one
Assay 96%
Molecular Formula C14H17N3O
MDL Number MFCD08061015
CAS 205058-28-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈235°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.