Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenyl-1,3-dioxane, 98%

$15.26 - $394.58

Chemical Identifiers

Chemical Name or Material 4-Phenyl-1,3-dioxane
CAS 772-00-9
Molecular Formula C10H12O2
Formula Weight 164.21g/mol
MDL Number MFCD00006568
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1583814

 
alfa aesar™
A15838-14
25g Each for $15.26

AAA1583822

 
alfa aesar™
A15838-22
100g Each for $47.41

AAA1583836

 
alfa aesar™
A15838-36
500g Each for $394.58
Description & Specifications

Specifications

Boiling Point 249° to 251°C
Quantity 25g
Flash Point >110°C (230°F)
Chemical Name or Material 4-Phenyl-1,3-dioxane
CAS 772-00-9
Assay 98%
Molecular Formula C10H12O2
Formula Weight 164.21g/mol
MDL Number MFCD00006568
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.