Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenyl-1,2,3-thiadiazole

$183.82 - $706.68

Chemical Identifiers

Chemical Name or Material 4-Phenyl-1,2,3-thiadiazole
CAS 25445-77-6
Molecular Formula C8H6N2S
Formula Weight 162.21
MDL Number MFCD00087232
Beilstein 119480
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2019703

 
alfa aesar™
L2019703
1g Each for $183.82

AAL2019706

 
alfa aesar™
L2019706
5g Each for $706.68
Description & Specifications

Specifications

Formula Weight 162.21
Chemical Name or Material 4-Phenyl-1,2,3-thiadiazole
Molecular Formula C8H6N2S
Quantity 1g
Melting Point 76° to 80°C
CAS 25445-77-6
MDL Number MFCD00087232
Beilstein 119480
Boiling Point ≈100°C (1.5mmHg)