missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Phenyl-1,2,3-thiadiazol-5-yl)methanol, 97%, Maybridge

Chemical Identifiers

CAS 423768-62-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H8N2OS
Formula Weight 192.24
MDL Number MFCD03086137
Chemical Name or Material (4-Phenyl-1, 2, 3-thiadiazol-5-yl)methanol
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC21509CB SDS Fisher Scientific Company
CC21509CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

423768-62-1
100.0
C9H8N2OS
MFCD03086137
97%
96.5
97%
192.24
(4-Phenyl-1, 2, 3-thiadiazol-5-yl)methanol

Specifications

192.24
100.0
96.5
97%
(4-Phenyl-1, 2, 3-thiadiazol-5-yl)methanol
250mg
C9H8N2OS
MFCD03086137
423768-62-1
97%