Please login to your online account to display your discounted pricing

(4-Phenyl-1,2,3-thiadiazol-5-yl)methanol, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  CC21509DA

 View more versions of this product

Catalog No. CC21509DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 192.24
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material (4-Phenyl-1, 2, 3-thiadiazol-5-yl)methanol
Assay Percent Range 97%
Molecular Formula C9H8N2OS
MDL Number MFCD03086137
CAS 423768-62-1
CAS Min % 96.5

Specifications

Chemical Name or Material (4-Phenyl-1, 2, 3-thiadiazol-5-yl)methanol
CAS 423768-62-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8N2OS
Formula Weight 192.24
MDL Number MFCD03086137