missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 99%, ACROS Organics™

Manufacturer:  Acros Organics 158860250

 View more versions of this product

Catalog No. AC158860250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride
Glass bottle
Brown
25g
43064-12-6
100.0
C11H14ClN
20, 3239
POGWXTJNUCZEPR-UHFFFAOYSA-N
4-phenyl-1,2,3,6-tetrahydropyridine;hydrochloride
2723860
Powder
Authentic
0.2% max. (K.F.)
200°C to 204°C
99%
98.5
99%
MFCD00012752
4-phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 1,2,3,6-tetrahydro-4-phenylpyridine hydrochloride, 4-phenyl-1,2,3,6-tetrahydroxpyridine, pyridine, 1,2,3,6-tetrahydro-4-phenyl-, hydrochloride, 4-phenyl-1,2,5,6-tetrahydropyridine, chloride, acmc-209jsh, 4-phenyl-1,2,3,6-tetra-hydropyridine, 4-phenyl-1,2,3,6-tetrahydropyridine hcl, 4-phenyl-1,2,3,6-tetrahydropyridinehydrochloride, 4-phenyl-1,2,3,6-tetrahydro-pyridine hydrochloride
C1CNCC=C1C2=CC=CC=C2.Cl
195.69
195.69
98 to 101% Total Base (on dry substance), ≥98.5% (TLC)

Chemical Identifiers

43064-12-6
195.69
POGWXTJNUCZEPR-UHFFFAOYSA-N
2723860
C1CNCC=C1C2=CC=CC=C2.Cl
C11H14ClN
MFCD00012752
4-phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 1,2,3,6-tetrahydro-4-phenylpyridine hydrochloride, 4-phenyl-1,2,3,6-tetrahydroxpyridine, pyridine, 1,2,3,6-tetrahydro-4-phenyl-, hydrochloride, 4-phenyl-1,2,5,6-tetrahydropyridine, chloride, acmc-209jsh, 4-phenyl-1,2,3,6-tetra-hydropyridine, 4-phenyl-1,2,3,6-tetrahydropyridine hcl, 4-phenyl-1,2,3,6-tetrahydropyridinehydrochloride, 4-phenyl-1,2,3,6-tetrahydro-pyridine hydrochloride
4-phenyl-1,2,3,6-tetrahydropyridine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 256-071-5

SDS Documents