missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 99%, ACROS Organics™

$537.85 - $537.85

Chemical Identifiers

CAS 43064-12-6
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C11H13N·HCl
Formula Weight 195.69
MDL Number MFCD00012752
Chemical Name or Material 4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 99%
Assay 98 to 101% Total Base (on dry substance), ≥98.5% (TLC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC158860100 SDS Thermo Fisher Scientific
158860100
Glass bottle 10g Each for $537.85
AC158860250 SDS Thermo Fisher Scientific
158860250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

43064-12-6
100.0
C11H13N·HCl
MFCD00012752
98 to 101% Total Base (on dry substance), ≥98.5% (TLC)
98.5
99%
195.69
4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 99%

Specifications

195.69
10g
20, 3239
100.0
Glass bottle
0.2% max. (K.F.)
43064-12-6
MFCD00012752
98 to 101% Total Base (on dry substance), ≥98.5% (TLC)
Powder
Light brown
200° to 204°C
98.5
Authentic
99%
C11H13N·HCl
4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 99%

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 256-071-5