Please login to your online account to display your discounted pricing

4-Phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride, Tocris

DA release inhibitor

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material 4-Phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Assay Percent Range >98%
Molecular Formula C15H15N·HCl
Formula Weight 245.75
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

073010

 
r & d systems
RDS073010
10mg Each for $177.37

073050

 
r & d systems
RDS073050
50mg Each for $746.48
Description & Specifications

Specifications

Chemical Name or Material 4-Phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Quantity 10mg
CAS 6109-35-9
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C15H15N·HCl
Formula Weight 245.75
Safety and Handling

Store at Room Temperature