Please login to your online account to display your discounted pricing

4-Phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride, Tocris


DA release inhibitor

Quantity 50mg

Manufacturer: r & d systems  RDS073050

 View more versions of this product

Catalog No. 073050

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 245.75
Chemical Name or Material 4-Phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Assay Percent Range >98%
Molecular Formula C15H15N·HCl
Recommended Storage Store at Room Temperature
CAS 6109-35-9

Specifications

Chemical Name or Material 4-Phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Assay Percent Range >98%
Molecular Formula C15H15N·HCl
Formula Weight 245.75
Accessories

Store at Room Temperature