Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenoxyphenylacetonitrile, 98%

$92.53 - $308.89

Chemical Identifiers

Chemical Name or Material 4-Phenoxyphenylacetonitrile
CAS 92163-15-0
Molecular Formula C14H11NO
Formula Weight 209.25
MDL Number MFCD00016399
Beilstein 2693571
Synonym 4-Phenoxybenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1057203

  SDS
alfa aesar™
L1057203
1g Each for $92.53

AAL1057206

  SDS
alfa aesar™
L1057206
5g Each for $308.89
Description & Specifications

Specifications

Formula Weight 209.25
Chemical Name or Material 4-Phenoxyphenylacetonitrile
MDL Number MFCD00016399
Health Hazard 1 Warning
Assay 98%
CAS 92163-15-0
Synonym 4-Phenoxybenzyl cyanide
Quantity 1g
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Molecular Formula C14H11NO
Beilstein 2693571
Boiling Point 145°C (1mmHg)

Applications
4-Phenoxyphenylacetonitrile is involved in the preparation of 2-(4-phenoxybenzoyl)-5-hydroxyindole, which is used as aCa2+/calmodulin-dependent protein kinase II (CaMKII) inhibitor.

Solubility
Miscible with toluene.

Notes
Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.