Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenoxyphenylacetic acid, 97%

$76.99 - $277.24

Chemical Identifiers

Chemical Name or Material 4-Phenoxyphenylacetic acid
CAS 6328-74-1
Molecular Formula C14H12O3
Formula Weight 228.25
MDL Number MFCD00079779
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2688603

  SDS
alfa aesar™
H2688603
1g Each for $76.99

AAH2688606

  SDS
alfa aesar™
H2688606
5g Each for $277.24
Description & Specifications

Specifications

Formula Weight 228.25
Molecular Formula C14H12O3
Melting Point 72° to 79°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Phenoxyphenylacetic acid
Health Hazard 1 Warning
CAS 6328-74-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00079779
Assay 97%
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.