Please login to your online account to display your discounted pricing

(4-Phenoxyphenyl)methylamine hydrochloride, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC53763DE

 View more versions of this product

Catalog No. CC53763DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 235.71
Quantity 5g
Assay 97%
CAS 169944-04-1
Chemical Name or Material (4-Phenoxyphenyl)methylamine hydrochloride
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C13H13NO·ClH
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material (4-Phenoxyphenyl)methylamine hydrochloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H13NO·ClH
Formula Weight 235.71
Accessories

GHS Signal Word: Warning