missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-phenoxyphenyl)methanol, 97%, Maybridge

$72.80 - $72.80

Chemical Identifiers

CAS 2215-78-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C13H12O2
Formula Weight 200.24
MDL Number MFCD01463970
Chemical Name or Material (4-Phenoxyphenyl)methanol
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC53709DA SDS Fisher Scientific Company
CC53709DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC53709CB SDS Fisher Scientific Company
CC53709CB
250mg Each for $72.80
Specifications Description & Specifications

Chemical Identifiers

2215-78-3
100.0
C13H12O2
MFCD01463970
97%
96.5
97%
200.24
(4-Phenoxyphenyl)methanol

Specifications

1g
96.5
Amber glass bottle
C13H12O2
MFCD01463970
97%
2215-78-3
100.0
97%
200.24
(4-Phenoxyphenyl)methanol

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.