missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenoxyphenyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L1168606

 View more versions of this product

Catalog No. AAL1168606



Specifications

Specifications

4-Phenoxyphenyl isocyanate
169°C to 170°C (3mmHg)
59377-19-4
MFCD00013876
393196
PNBUGOFIKAHZRW-UHFFFAOYSA-N
1-isocyanato-4-phenoxybenzene
2734896
98%
>110°C (230°F)
5g
C13H9NO2
UN2206
4-phenoxyphenyl isocyanate, 4-phenoxyphenylisocyanate, 1-isocyanato-4-phenoxy-benzene, benzene, 1-isocyanato-4-phenoxy, acmc-20ambk, pubchem17221, p-phenoxyphenylisocyanate, 4-phenoxybenzenisocyanate, 4-phenoxyphenyl-isocyanate, 4-isocyanatodiphenyl ether
C1=CC=C(C=C1)OC2=CC=C(C=C2)N=C=O
211.22
211.22

Chemical Identifiers

59377-19-4
211.22
PNBUGOFIKAHZRW-UHFFFAOYSA-N
2734896
C1=CC=C(C=C1)OC2=CC=C(C=C2)N=C=O
C13H9NO2
MFCD00013876
4-phenoxyphenyl isocyanate, 4-phenoxyphenylisocyanate, 1-isocyanato-4-phenoxy-benzene, benzene, 1-isocyanato-4-phenoxy, acmc-20ambk, pubchem17221, p-phenoxyphenylisocyanate, 4-phenoxybenzenisocyanate, 4-phenoxyphenyl-isocyanate, 4-isocyanatodiphenyl ether
1-isocyanato-4-phenoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents