Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenoxyphenol, 99%

$73.12 - $577.23

Chemical Identifiers

Chemical Name or Material 4-Phenoxyphenol
CAS 831-82-3
Molecular Formula C12H10O2
Formula Weight 186.21
MDL Number MFCD00002331
Beilstein 2047182
Synonym Hydroquinone monophenyl ether; 4-Hydroxydiphenyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1848906

  SDS
alfa aesar™
A1848906
5g Each for $73.12

AAA1848914

  SDS
alfa aesar™
A1848914
25g Each for $209.34

AAA1848922

  SDS
alfa aesar™
A1848922
100g Each for $577.23
Description & Specifications

Specifications

Formula Weight 186.21
Chemical Name or Material 4-Phenoxyphenol
TSCA TSCA
Assay 99%
Molecular Formula C12H10O2
MDL Number MFCD00002331
CAS 831-82-3
Beilstein 2047182
Synonym Hydroquinone monophenyl ether; 4-Hydroxydiphenyl ether
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 170°C (338°F)
Boiling Point 177° to 180°C (9mmHg)
Melting Point 83° to 85°C
Quantity 5g

Applications
4-Phenoxyphenol is a part of a group of phenolic compounds that have potential apoptotic activity, which can be used in the development of new treatments for cancer therapy.

Solubility
Soluble in water (partly).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.