Please login to your online account to display your discounted pricing

4-Phenoxyphenethylamine, +97%, ACROS Organics™


Quantity 250mg
Packaging Glass Ampule with screw-cap

Manufacturer: thermo fisher scientific  AC393492500

 View more versions of this product

Catalog No. AC393492500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.0%
Physical Form Liquid
Formula Weight 213.28
Quantity 250mg
Color Clear yellow
Density 1.09g/mL
Beilstein 13, IV, 1789
CAS Min % 97.0
CAS Max % 100.0
Chemical Name or Material 4-Phenoxyphenethylamine
Packaging Glass Ampule with screw-cap
Refractive Index 1.5830 to 1.5850 (20°C, 589nm)
Assay Percent Range 97+%
Molecular Formula C14H15NO
Linear Formula C6H5OC6H4CH2CH2NH2
MDL Number MFCD00079777
CAS 118468-18-1
Synonym 4-Aminoethylphenyl phenyl ether
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 4-Phenoxyphenethylamine
CAS 118468-18-1
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C14H15NO
Linear Formula C6H5OC6H4CH2CH2NH2
Formula Weight 213.28
MDL Number MFCD00079777
Synonym 4-Aminoethylphenyl phenyl ether
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents