missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Phenoxyphenethylamine, +97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC393490010

 View more versions of this product

Catalog No. AC393490010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.0%
213.28
Clear yellow
1.09g/mL
97.0
C6H5OC6H4CH2CH2NH2
118468-18-1
97+%
Glass bottle
4-Phenoxyphenethylamine
Liquid
1g
13, IV, 1789
100.0
4-Aminoethylphenyl phenyl ether
1.5830 to 1.5850 (20°C, 589nm)
C14H15NO
MFCD00079777
Authentic

Chemical Identifiers

118468-18-1
100.0
C14H15NO
213.28
4-Aminoethylphenyl phenyl ether
≥97.0%
97.0
97+%
C6H5OC6H4CH2CH2NH2
MFCD00079777
4-Phenoxyphenethylamine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger