Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenoxybutyryl chloride, 97%

Chemical Identifiers

Chemical Name or Material 4-Phenoxybutyryl chloride
CAS 5139-89-9
Molecular Formula C10H11ClO2
Linear Formula C6H5O(CH2)3COCl
Formula Weight 198.65
MDL Number MFCD02094040
UN Number UN3265
Beilstein 2102483
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0313006

 
alfa aesar™
L0313006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198.65
Chemical Name or Material 4-Phenoxybutyryl chloride
MDL Number MFCD02094040
Assay 97%
Molecular Formula C10H11ClO2
Linear Formula C6H5O(CH2)3COCl
CAS 5139-89-9
UN Number UN3265
Beilstein 2102483
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Boiling Point 154° to 156°C (20mmHg)
Flash Point 111°C (232°F)
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 5g
Odor Irritating
Health Hazard 1 Danger