Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenoxybutyric acid, 99%

$91.73 - $91.73

Chemical Identifiers

Chemical Name or Material 4-Phenoxybutyric acid
CAS 6303-58-8
Molecular Formula C6H5O(CH2)3CO2H
Formula Weight 180.21
MDL Number MFCD00042656
Beilstein 1640610
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0194914

 
alfa aesar™
L0194914
25g Each for $91.73
Description & Specifications

Specifications

Formula Weight 180.21
Chemical Name or Material 4-Phenoxybutyric acid
Molecular Formula C6H5O(CH2)3CO2H
MDL Number MFCD00042656
CAS 6303-58-8
Assay 99%
Beilstein 1640610
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number ET5956300
Melting Point 62° to 64°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.