Please login to your online account to display your discounted pricing

4-Phenoxybenzenesulfonyl chloride, 97%, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  AC450350050

 View more versions of this product

Catalog No. AC450350050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 4-Phenoxybenzenesulfonyl chloride
Quantity 5g
CAS 1623-92-3
Assay Percent Range 97%
Molecular Formula C12H9ClO3S
Formula Weight 268.72

Specifications

Chemical Name or Material 4-Phenoxybenzenesulfonyl chloride
Assay Percent Range 97%
Molecular Formula C12H9ClO3S
Formula Weight 268.72