missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Phenoxybenzenesulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450350010

 View more versions of this product

Catalog No. AC450350010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Phenoxybenzenesulfonyl chloride
43.0°C to 45.0°C
1623-92-3
C12H9ClO3S
QIZPONOMFWAPRR-UHFFFAOYSA-N
4-phenoxybenzenesulfonyl chloride
2794756
97%
Glass Bottle
1g
96% min. diethylsulfonamide derivative (HPLC)
4-phenoxy benzene sulfonyl chloride, 4-phenoxy-benzenesulfonyl chloride, 4-phenoxy-benzenesulphonyl chloride, 4-phenoxybenzene-1-sulfonyl chloride, 4-phenoxybenzene-sulfonyl chloride, 4-phenoxybenzenesulphonyl chloride, 4-phenoxyphenylsulfonyl chloride, acmc-1bvtb, benzenesulfonyl chloride, 4-phenoxy, chloro 4-phenoxyphenyl sulfone
C1=CC=C(C=C1)OC2=CC=C(C=C2)S(=O)(=O)Cl
268.72
268.72

Chemical Identifiers

1623-92-3
268.72
4-phenoxy benzene sulfonyl chloride, 4-phenoxy-benzenesulfonyl chloride, 4-phenoxy-benzenesulphonyl chloride, 4-phenoxybenzene-1-sulfonyl chloride, 4-phenoxybenzene-sulfonyl chloride, 4-phenoxybenzenesulphonyl chloride, 4-phenoxyphenylsulfonyl chloride, acmc-1bvtb, benzenesulfonyl chloride, 4-phenoxy, chloro 4-phenoxyphenyl sulfone
4-phenoxybenzenesulfonyl chloride
C12H9ClO3S
QIZPONOMFWAPRR-UHFFFAOYSA-N
2794756
C1=CC=C(C=C1)OC2=CC=C(C=C2)S(=O)(=O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger

SDS Documents