Please login to your online account to display your discounted pricing

4-Phenoxybenzenesulfonyl chloride, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-Phenoxybenzenesulfonyl chloride
Assay Percent Range 97%
Molecular Formula C12H9ClO3S
Formula Weight 268.72
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC450350010

  SDS
thermo fisher scientific
AC450350010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC450350050

 
thermo fisher scientific
AC450350050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 268.72
Quantity 1g
CAS 1623-92-3
Assay Percent Range 97%
Molecular Formula C12H9ClO3S
Chemical Name or Material 4-Phenoxybenzenesulfonyl chloride