Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenoxybenzenesulfonamide, 98%

Chemical Identifiers

Chemical Name or Material 4-Phenoxybenzenesulfonamide
CAS 123045-62-5
Molecular Formula C12H11NO3S
Formula Weight 249.29
MDL Number MFCD00623227
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27486MD

  SDS
alfa aesar™
H27486MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.29
Quantity 250mg
Chemical Name or Material 4-Phenoxybenzenesulfonamide
Melting Point 130° to 132°C
MDL Number MFCD00623227
CAS 123045-62-5
Assay 98%
Molecular Formula C12H11NO3S