Learn More
4-Phenoxyaniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 130231000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Phenoxyaniline | |
96.0 | |
82°C to 86°C | |
140°C (3.0 hPa) | |
97% | |
C12H11NO | |
MFCD00007862 | |
13, 438 | |
Solubility in water: < 1g/l in water (20°C). Other solubilities: soluble in isopropanol,toluene,very soluble in acetone,1,2-dichloroethane | |
C1=CC=C(C=C1)OC2=CC=C(C=C2)N | |
185.23 | |
185.23 | |
Crystalline Flakes or Powder |
139-59-3 | |
100.0 | |
Brown to Brown-Green | |
Authentic | |
Glass bottle | |
C6H5OC6H4NH2 | |
100 g | |
p-phenoxyaniline, 4-aminodiphenyl ether, benzenamine, 4-phenoxy, 4-phenoxybenzenamine, 4-aminophenyl phenyl ether, 4-aminodifenylether, 4-amino-1-phenoxybenzene, aniline, p-phenoxy, 4-aminobiphenyl ether, 4-aminodiphenylether | |
WOYZXEVUWXQVNV-UHFFFAOYSA-N | |
4-phenoxyaniline | |
8764 | |
97% |
Chemical Identifiers
139-59-3 | |
WOYZXEVUWXQVNV-UHFFFAOYSA-N | |
8764 | |
C1=CC=C(C=C1)OC2=CC=C(C=C2)N |
MFCD00007862 | |
p-phenoxyaniline, 4-aminodiphenyl ether, benzenamine, 4-phenoxy, 4-phenoxybenzenamine, 4-aminophenyl phenyl ether, 4-aminodifenylether, 4-amino-1-phenoxybenzene, aniline, p-phenoxy, 4-aminobiphenyl ether, 4-aminodiphenylether | |
4-phenoxyaniline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF IN EYES: Rinse
GHS Signal Word: Warning
EINECSNumber : 205-367-2
RUO – Research Use Only