Please login to your online account to display your discounted pricing

4'-Phenoxyacetophenone, 98%, ACROS Organics™

$73.66 - $73.66

Chemical Identifiers

Chemical Name or Material 4'-Phenoxyacetophenone
Name Note 98%
CAS 5031-78-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H12O2
Formula Weight 212.25
MDL Number MFCD00008744
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334980250

  SDS
thermo fisher scientific
334980250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC334980050

  SDS
thermo fisher scientific
334980050
Glass bottle 5g Each for $73.66
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 212.25
Quantity 25g
Color Light yellow
TSCA TSCA
Melting Point 48° to 52°C
CAS Max % 100.0
Chemical Name or Material 4'-Phenoxyacetophenone
Infrared Spectrum Authentic
Name Note 98%
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C14H12O2
MDL Number MFCD00008744
CAS 5031-78-7
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.