missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Phenoxyacetophenone, 98+%

Manufacturer:  Alfa Aesar™ A1239214

 View more versions of this product

Catalog No. AAA1239214



Specifications

Specifications

4'-Phenoxyacetophenone
5031-78-7
C14H12O2
2049295
DJNIFZYQFLFGDT-UHFFFAOYSA-N
1-(4-phenoxyphenyl)ethanone
236783
325°C
>110°C (230°F)
25g
≥98%
MFCD00008744
4-Acetyldiphenyl ether
CC(=O)C1=CC=C(C=C1)OC2=CC=CC=C2
212.248
212.25
48°C to 52°C

Chemical Identifiers

5031-78-7
212.248
DJNIFZYQFLFGDT-UHFFFAOYSA-N
236783
CC(=O)C1=CC=C(C=C1)OC2=CC=CC=C2
C14H12O2
MFCD00008744
4-Acetyldiphenyl ether
1-(4-phenoxyphenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents