Please login to your online account to display your discounted pricing

4-Phenolsulfonic Acid, Sodium Salt Dihydrate 99%, ACROS Organics™

$35.64 - $107.78

Chemical Identifiers

Chemical Name or Material 4-Phenolsulfonic acid, sodium salt dihydrate
Name Note 99%
CAS 10580-19-5
CAS Min % 98.5
CAS Max % 100.0
CAS 825-90-1
Assay ≥98.5% (Acid)
Assay Percent Range 99%
Molecular Formula C6H5NaO4S·2H2O
Linear Formula HOC6H4SO3Na·2H2O
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC123980050

  SDS
thermo fisher scientific
AC123980050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC123981000

  SDS
fisher scientific acros
123981000
Plastic bottle 100g Each for $35.64

AC123985000

  SDS
thermo fisher scientific
123985000
Plastic bottle 500g Each for $107.78

AC123980025

  SDS
thermo fisher scientific
AC123980025
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5% (Acid)
Physical Form Powder
Formula Weight 232.18
Quantity 5g
Color Pale yellow to beige to gold to green
EINECS Number 212-550-0
TSCA TSCA
Water 13 to 17% (K.F.)
CAS Max % 100.0
Infrared Spectrum Authentic
Name Note 99%
Packaging Glass bottle
Assay Percent Range 99%
MDL Number MFCD00150724
CAS 10580-19-5
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
CAS 825-90-1
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Phenolsulfonic acid, sodium salt dihydrate
Molecular Formula C6H5NaO4S·2H2O
Linear Formula HOC6H4SO3Na·2H2O
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.