Learn More
4-Pentyn-1-ol, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1040522
Description
4-Pentyn-1-ol is widely utilized in ring-opening polymerization reactions. It plays an important role as a starting reagent in stereo selective total synthesis of antimicrobial marine metabolites, ieodomycin A and B. It is also used in the preparation of 3-pent-4-ynyloxy phthalonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Pentyn-1-ol is widely utilized in ring-opening polymerization reactions. It plays an important role as a starting reagent in stereo selective total synthesis of antimicrobial marine metabolites, ieodomycin A and B. It is also used in the preparation of 3-pent-4-ynyloxy phthalonitrile.
Solubility
Miscible with water.
Notes
Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
4-Pentyn-1-ol | |
0.907 | |
62°C (143°F) | |
1.445 | |
MFCD00002974 | |
1736712 | |
Miscible with water. | |
C#CCCCO | |
84.12 | |
84.12 |
5390-04-5 | |
153°C to 155°C | |
C5H8O | |
HC≡C(CH2)3OH | |
100 g | |
4-pentyn-1-ol, 4-pentyne-1-ol, 1-pentyn-5-ol, 5-hydroxypentyne, 4-pentynol, 4-pentin-1-ol, pent-4yn-1-ol, 5-hydroxy-1-pentyne, pent-1-yn-5-ol, pubchem2025 | |
CRWVOXFUXPYTRK-UHFFFAOYSA-N | |
pent-4-yn-1-ol | |
79346 | |
98% |
Chemical Identifiers
5390-04-5 | |
MFCD00002974 |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
EINECSNumber : 226-383-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only