missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Pentylbenzonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB12834EA

 View more versions of this product

Catalog No. BTB12834EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Pentylbenzonitrile
96.5
97%
C12H15N
MFCD00218832
4-n-pentylbenzonitrile, 4-pentylbenzenecarbonitrile, p-amylbenzonitrile, p-n-pentylbenzonitrile, benzonitrile, 4-pentyl
CCCCCC1=CC=C(C=C1)C#N
173.26
173.26
10270-29-8
100.0
Amber Glass Bottle
CH3(CH2)4C6H4CN
10g
LGPQFJXPPKNHJU-UHFFFAOYSA-N
4-pentylbenzonitrile
264951
97%

Chemical Identifiers

10270-29-8
173.26
LGPQFJXPPKNHJU-UHFFFAOYSA-N
264951
CCCCCC1=CC=C(C=C1)C#N
C12H15N
MFCD00218832
4-n-pentylbenzonitrile, 4-pentylbenzenecarbonitrile, p-amylbenzonitrile, p-n-pentylbenzonitrile, benzonitrile, 4-pentyl
4-pentylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions