Learn More
4-Pentylbenzoic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals BTB09863EA
Specifications
4-Pentylbenzoic acid | |
96.5 | |
97% | |
CH3(CH2)4C6H4CO2H | |
10g | |
CWYNKKGQJYAHQG-UHFFFAOYSA-N | |
4-pentylbenzoic acid | |
33479 | |
97% |
26311-45-5 | |
100.0 | |
C12H16O2 | |
MFCD00002572 | |
benzoic acid, 4-pentyl, 4-n-pentylbenzoic acid, p-pentylbenzoic acid, 4-amylbenzoic acid, p-amylbenzoic acid, para-pentylbenzoic acid, benzoic acid, p-pentyl, 4-pentyl benzoic acid, p-n-pentylbenzoic acid, 4-pentylbenzoicacid | |
CCCCCC1=CC=C(C=C1)C(=O)O | |
192.258 | |
192.26 |
Chemical Identifiers
26311-45-5 | |
192.258 | |
CWYNKKGQJYAHQG-UHFFFAOYSA-N | |
33479 | |
CCCCCC1=CC=C(C=C1)C(=O)O |
C12H16O2 | |
MFCD00002572 | |
benzoic acid, 4-pentyl, 4-n-pentylbenzoic acid, p-pentylbenzoic acid, 4-amylbenzoic acid, p-amylbenzoic acid, para-pentylbenzoic acid, benzoic acid, p-pentyl, 4-pentyl benzoic acid, p-n-pentylbenzoic acid, 4-pentylbenzoicacid | |
4-pentylbenzoic acid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.